Learn More
Abnova™ Human USP28 Partial ORF (NP_065937, 978 a.a. - 1077 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057646-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The ubiquitin-dependent protein degradation pathway is essential for proteolysis of intracellular proteins and peptides. Enzymes that remove ubiquitin from ubiquitin-conjugated peptides, like USP28, affect the fate and degradation of intracellular proteins and are essential for maintenance of cell-free ubiquitin pools (Valero et al., 2001).[supplied by OMIM]
Sequence: VMNELIIPCIHLIINNDISKDDLDAIEVMRNHWCSYLGQDIAENLQLCLGEFLPRLLDPSAEIIVLKEPPTIRPNSPYDLCSRFAAVMESIQGVSTVTVKEspecificaciones
NP_065937 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VMNELIIPCIHLIINNDISKDDLDAIEVMRNHWCSYLGQDIAENLQLCLGEFLPRLLDPSAEIIVLKEPPTIRPNSPYDLCSRFAAVMESIQGVSTVTVK | |
RUO | |
USP28 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57646 | |
USP28 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1515 | |
USP28 | |
Recombinant | |
wheat germ expression system |