missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UNR Partial ORF (NP_009089, 670 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007812-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16197965

  • 302.00€ / 10 microgramos


Fecha estimada de envío
23-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: HVKEVQDGIELQAGDEVEFSVILNQRTGKCSACNVWRVCEGPKAVAAPRPDRLVNRLKNITLDDASAPRLMVLRQPRGPDNSMGFGAERKIRQAGV
Especificaciones

Especificaciones

NP_009089
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HVKEVQDGIELQAGDEVEFSVILNQRTGKCSACNVWRVCEGPKAVAAPRPDRLVNRLKNITLDDASAPRLMVLRQPRGPDNSMGFGAERKIRQAGV
RUO
CSDE1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
7812
UNR (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D1S155E|DKFZp779B0247|DKFZp779J1455|FLJ26882|RP5-1000E10.3|UNR
CSDE1
Yes
Videos
Documentos