Learn More
Abnova™ Human TSNAX Full-length ORF (NP_005990.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007257-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein which specifically interacts with translin, a DNA-binding protein that binds consensus sequences at breakpoint junctions of chromosomal translocations. The encoded protein contains bipartite nuclear targeting sequences that may provide nuclear transport for translin, which lacks any nuclear targeting motifs. [provided by RefSeq]
Sequence: MSNKEGSGGFRKRKHDNFPHNQRREGKDVNSSSPVMLAFKSFQQELDARHDKYERLVKLSRDITVESKRTIFLLHRITSAPDMEDILTESEIKLDGVRQKIFQVAQELSGEDMHQFHRAITTGLQEYVEAVSFQHFIKTRSLISMDEINKQLIFTTEDNGKENKTPSSDAQDKQFGTWRLRVTPVDYLLGVADLTGELMRMCINSVGNGDIDTPFEVSQFLRQVYDGFSFIGNTGPYEVSKKLYTLKQSLAKVENACYALKVRGSEIPKHMLADVFSVKTEMIDQEEGISEspecificaciones
NP_005990.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TRAX | |
TSNAX | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7257 | |
TSNAX (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSNKEGSGGFRKRKHDNFPHNQRREGKDVNSSSPVMLAFKSFQQELDARHDKYERLVKLSRDITVESKRTIFLLHRITSAPDMEDILTESEIKLDGVRQKIFQVAQELSGEDMHQFHRAITTGLQEYVEAVSFQHFIKTRSLISMDEINKQLIFTTEDNGKENKTPSSDAQDKQFGTWRLRVTPVDYLLGVADLTGELMRMCINSVGNGDIDTPFEVSQFLRQVYDGFSFIGNTGPYEVSKKLYTLKQSLAKVENACYALKVRGSEIPKHMLADVFSVKTEMIDQEEGIS | |
RUO | |
TSNAX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |