missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSKS Full-length ORF (AAH58862.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00060385-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16103193

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 30-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression. [provided by RefSeq]

Sequence: MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA
Especificaciones

Especificaciones

AAH58862.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TSKS1
TSKS
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
60385
TSKS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA
RUO
TSKS
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales