missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRY1 Full-length ORF (NP_001001317.1, 1 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00136541-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198083

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the trypsin family of serine proteases. This gene and several related trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7. This gene was previously described as a trypsinogen-like pseudogene, but it is now thought to be a protein-coding gene. [provided by RefSeq]

Sequence: MKFILLWALLNLTVALAFNPDYTVSSTPPYLVYLKSDYLPCAGVLIHPLWVITAAHCNLPKLRVILGVTIPADSNEKHLQVIGYEKMIHHPHFSVTSIDHDIMLIKLKTEAELNDYVKLANLPYQTISENTMCSVSTWSYNVCDIYKEPDSLQTVNISVISKPQCRDAYKTYNITENMLCVGIVPGRRQPCKEVSAAPAICNGMLQGILSFADGCVLRADVGIYAKIFYYIPWIENVIQNN
Especificaciones

Especificaciones

NP_001001317.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16649/MGC35022/TRY1/UNQ2540
TRYX3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
136541
TRY1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKFILLWALLNLTVALAFNPDYTVSSTPPYLVYLKSDYLPCAGVLIHPLWVITAAHCNLPKLRVILGVTIPADSNEKHLQVIGYEKMIHHPHFSVTSIDHDIMLIKLKTEAELNDYVKLANLPYQTISENTMCSVSTWSYNVCDIYKEPDSLQTVNISVISKPQCRDAYKTYNITENMLCVGIVPGRRQPCKEVSAAPAICNGMLQGILSFADGCVLRADVGIYAKIFYYIPWIENVIQNN
RUO
TRYX3
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales