Learn More
Abnova™ Human TRPV5 Partial ORF (NP_062815, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056302-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level. [provided by RefSeq]
Sequence: MGGFLPKAEGPGSQLQKLLPSFLVREQDWDQHLDKLHMLQQKRILESPLLRASKENDLSVLRQLLLDCTCDVRQRGALGETALHIAALYDNLEAALVLMEEspecificaciones
NP_062815 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGFLPKAEGPGSQLQKLLPSFLVREQDWDQHLDKLHMLQQKRILESPLLRASKENDLSVLRQLLLDCTCDVRQRGALGETALHIAALYDNLEAALVLME | |
RUO | |
TRPV5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56302 | |
TRPV5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAT2/ECAC1/OTRPC3 | |
TRPV5 | |
Recombinant | |
wheat germ expression system |