missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRPC4AP Full-length ORF (NP_056453.1, 1 a.a. - 797 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

en la oferta

Especificaciones

Nombre TRPC4AP (Human) Recombinant Protein (P01)
Recombinante Yes
Método de purificación Glutathione Sepharose 4 Fast Flow
Peso molecular 117.3kDa
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ver más especificaciones

Descripción

Descripción

Sequence: MAAAPVAAGSGAGRGRRSAATVAAWGGWGGRPRPGNILLQLRQGQLTGRGLVRAVQFTETFLTERDKQSKWSGIPQLLLKLHTTSHLHSDFVECQNILKEISPLLSMEAMAFVTEERKLTQETTYPNTYIFDLFGGVDLLVEILMRPTISIRGQKLKISDEMSKDCLSILYNTCVCTEGVTKRLAEKNDFVIFLFTLMTSKKTFLQTATLIEDILGVKKEMIRLDEVPNLSSLVSNFDQQQLANFCRILAVTISEMDTGNDDKHTLLAKNAQQKKSLSLGPSAAEINQAALLSIPGFVERLCKLATRKVSESTGTASFLQELEEWYTWLDNALVLDALMRVANEESEHNQASIVFPPPGASEENGLPHTSARTQLPQSMKIMHEIMYKLEVLYVLCVLLMGRQRNQVHRMIAEFKLIPGLNNLFDKLIWRKHSASALVLHGHNQNCDCSPDITLKIQFLRLLQSFSDHHENKYLLLNNQELNELSAISLKANIPEVEAVLNTDRSLVCDGKRGLLTRLLQVMKKEPAESSFRFWQARAVESFLRGTTSYADQMFLLKRGLLEHILYCIVDSECKSRDVLQSYFDLLGELMKFNVDAFKRFNKYINTDAKFQVFLKQINSSLVDSNMLVRCVTLSLDRFENQVDMKVAEVLSECRLLAYISQVPTQMSFLFRLINIIHVQTLTQENVSCLNTSLVILMLARRKERLPLYLRLLQRMEHSKKYPGFLLNNFHNLLRFWQQHYLHKDKDSTCLENSSCISFSYWKETVSILLNPDRQSPSALVSYIEEPYMDIDRDFTEE
Especificaciones

Especificaciones

TRPC4AP (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
117.3kDa
Videos
SDS
Documentos

Documentos

Product Certifications