Learn More
Abnova™ Human TRIM26 Partial ORF (NP_003440, 262 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007726-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6. [provided by RefSeq]
Sequence: NRYPRKKFWVGKPIARVVKKKTGEFSDKLLSLQRGLREFQGKLLRDLEYKTVSVTLDPQSASGYLQLSEDWKCVTYTSLYKSAYLHPQQFDCEPGVLGSKGFTWGKVYWEspecificaciones
NP_003440 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NRYPRKKFWVGKPIARVVKKKTGEFSDKLLSLQRGLREFQGKLLRDLEYKTVSVTLDPQSASGYLQLSEDWKCVTYTSLYKSAYLHPQQFDCEPGVLGSKGFTWGKVYW | |
RUO | |
TRIM26 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7726 | |
TRIM26 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AFP/RNF95/ZNF173 | |
TRIM26 | |
Recombinant | |
wheat germ expression system |