missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM23 Full-length ORF (AAH22510, 1 a.a. - 574 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000373-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16101781

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein is also a member of the ADP ribosylation factor family of guanine nucleotide-binding family of proteins. Its carboxy terminus contains an ADP-ribosylation factor domain and a guanine nucleotide binding site, while the amino terminus contains a GTPase activating protein domain which acts on the guanine nucleotide binding site. The protein localizes to lysosomes and the Golgi apparatus. It plays a role in the formation of intracellular transport vesicles, their movement from one compartment to another, and phopholipase D activation. Three alternatively spliced transcript variants for this gene have been described. [provided by RefSeq]

Sequence: MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQYGAAEESIGISGESIIRCDEDEAHLASVYCTVCATHLCSECSQVTHSTKTLAKHRRVPLADKPHEKTMCSQHQVHAIEFVCLEEGCQTSPLMCCVCKEYGKHQGHKHSVLEPEANQIRASILDMAHCIRTFTEEISDYSRKLVGIVQHIEGGEQIVEDGIGMAHTEHVPGTAENARSCIRAYFYDLHETLCRQEEMALSVVDAHVREKLIWLRQQQEDMTILLSEVSAACLHCEKTLQQDDCRVVLAKQEITRLLETLQKQQQQFTEVADHIQLDASIPVTFTKDNRVHIGPKMEIRVVTLGLDGAGKTTILFKLKQDEFMQPIPTIGFNVETVEYKNLKFTIWDVGGKHKLRPLWKHYYLNTQAVVFVVDSSHRDRISEAHSELAKLLTEKELRDALLLIFANKQDVAGALSVEEITELLSLHKLCCGRSWYIQGCDARSGMGLYEGLDWLSRQLVAAGVLDVA
Especificaciones

Especificaciones

AAH22510
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.88kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARD1/ARFD1/RNF46
TRIM23
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
373
TRIM23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQYGAAEESIGISGESIIRCDEDEAHLASVYCTVCATHLCSECSQVTHSTKTLAKHRRVPLADKPHEKTMCSQHQVHAIEFVCLEEGCQTSPLMCCVCKEYGKHQGHKHSVLEPEANQIRASILDMAHCIRTFTEEISDYSRKLVGIVQHIEGGEQIVEDGIGMAHTEHVPGTAENARSCIRAYFYDLHETLCRQEEMALSVVDAHVREKLIWLRQQQEDMTILLSEVSAACLHCEKTLQQDDCRVVLAKQEITRLLETLQKQQQQFTEVADHIQLDASIPVTFTKDNRVHIGPKMEIRVVTLGLDGAGKTTILFKLKQDEFMQPIPTIGFNVETVEYKNLKFTIWDVGGKHKLRPLWKHYYLNTQAVVFVVDSSHRDRISEAHSELAKLLTEKELRDALLLIFANKQDVAGALSVEEITELLSLHKLCCGRSWYIQGCDARSGMGLYEGLDWLSRQLVAAGVLDVA
RUO
TRIM23
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales