Learn More
Abnova™ Human TRIM17 Partial ORF (NP_057186.1, 75 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051127-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: LPNRLLTKVAEMAQQHPGLQKQDLCQEHHEPLKLFCQKDQSPICVVCRESREHRLHRVLPAEEAVQGYKLKLEEDMEYLREQITRTGNLQAREEQSLAEWQGKVKERREREspecificaciones
NP_057186.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPNRLLTKVAEMAQQHPGLQKQDLCQEHHEPLKLFCQKDQSPICVVCRESREHRLHRVLPAEEAVQGYKLKLEEDMEYLREQITRTGNLQAREEQSLAEWQGKVKERRER | |
RUO | |
TRIM17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51127 | |
TRIM17 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBCC/RNF16/terf | |
TRIM17 | |
Recombinant | |
wheat germ expression system |