Learn More
Abnova™ Human TPM2 Full-length ORF (AAH11776, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007169-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes beta-tropomyosin, a member of the actin filament binding protein family, and mainly expressed in slow, type 1 muscle fibers. Mutations in this gene can alter the expression of other sarcomeric tropomyosin proteins, and cause cap disease, nemaline myopathy and distal arthrogryposis syndromes. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Sequence: MDAIKKKMQMLKLDKENAIDRAEQAEADKKQAEDRCKQLEEEQQALQKKLKGTEDEVEKYSESVKEAQEKLEQAEKKATDAEADVASLNRRIQLVEEELDRAQERLATALQKLEEAEKAADESERGMKVIENRAMKDEEKMELQEMQLKEAKHIAEDSDRKYEEVARKLVILEGELERSEERAEVAESRARQLEEELRTMDQALKSLMASEEEYSTKEDKYEEEIKLLEEKLKEAETRAEFAERSVAKLEKTIDDLEETLASAKEENVEIHQTLDQTLLELNNLEspecificaciones
AAH11776 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.98kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMCD1/DA1/DA2B/TMSB | |
TPM2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7169 | |
TPM2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDAIKKKMQMLKLDKENAIDRAEQAEADKKQAEDRCKQLEEEQQALQKKLKGTEDEVEKYSESVKEAQEKLEQAEKKATDAEADVASLNRRIQLVEEELDRAQERLATALQKLEEAEKAADESERGMKVIENRAMKDEEKMELQEMQLKEAKHIAEDSDRKYEEVARKLVILEGELERSEERAEVAESRARQLEEELRTMDQALKSLMASEEEYSTKEDKYEEEIKLLEEKLKEAETRAEFAERSVAKLEKTIDDLEETLASAKEENVEIHQTLDQTLLELNNL | |
RUO | |
TPM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |