missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TOB1 Full-length ORF (NP_005740.1, 1 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010140-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16194552

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90 kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells. [provided by RefSeq]

Sequence: MQLEIQVALNFIISYLYNKLPRRRVNIFGEELERLLKKKYEGHWYPEKPYKGSGFRCIHIGEKVDPVIEQASKESGLDIDDVRGNLPQDLSVWIDPFEVSYQIGEKGPVKVLYVDDNNENGCELDKEIKNSFNPEAQVFMPISDPASSVSSSPSPPFGHSAAVSPTFMPRSTQPLTFTTATFAATKFGSTKMKNSGRSNKVARTSPINLGLNVNDLLKQKAISSSMHSLYGLGLGSQQQPQQQQQPAQPPPPPPPPQQQQQQKTSALSPNAKEFIFPNMQGQGSSTNGMFPGDSPLNLSPLQYSNAFDVFAAYGGLNEKSFVDGLNFSLNNMQYSNQQFQPVMAN
Especificaciones

Especificaciones

NP_005740.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APRO6/MGC104792/MGC34446/PIG49/TOB/TROB/TROB1
TOB1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10140
TOB1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQLEIQVALNFIISYLYNKLPRRRVNIFGEELERLLKKKYEGHWYPEKPYKGSGFRCIHIGEKVDPVIEQASKESGLDIDDVRGNLPQDLSVWIDPFEVSYQIGEKGPVKVLYVDDNNENGCELDKEIKNSFNPEAQVFMPISDPASSVSSSPSPPFGHSAAVSPTFMPRSTQPLTFTTATFAATKFGSTKMKNSGRSNKVARTSPINLGLNVNDLLKQKAISSSMHSLYGLGLGSQQQPQQQQQPAQPPPPPPPPQQQQQQKTSALSPNAKEFIFPNMQGQGSSTNGMFPGDSPLNLSPLQYSNAFDVFAAYGGLNEKSFVDGLNFSLNNMQYSNQQFQPVMAN
RUO
TOB1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales