missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TMSB4X Full-length ORF (NP_066932, 1 a.a. - 44 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007114-P02.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191452

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y. [provided by RefSeq]

Sequence: MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGES
Especificaciones

Especificaciones

NP_066932
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.58kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FX/PTMB4/TB4X/TMSB4
TMSB4X
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7114
TMSB4X (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGES
RUO
TMSB4X
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales