Learn More
Abnova™ Human TMIE Partial ORF (NP_671729, 79 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00259236-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a transmembrane inner ear protein. Studies in mouse suggest that this gene is required for normal postnatal maturation of sensory hair cells in the cochlea, including correct development of stereocilia bundles. This gene is one of multiple genes responsible for recessive non-syndromic deafness (DFNB), also known as autosomal recessive nonsyndromic hearing loss (ARNSHL), the most common form of congenitally acquired inherited hearing impairment. [provided by RefSeq]
Sequence: NCRVPRTRKEIEARYLQRKAAKMYTDKLETVPPLNELTEVPGEDKKKKKKKKDSVDTVAIKVEspecificaciones
NP_671729 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.56kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NCRVPRTRKEIEARYLQRKAAKMYTDKLETVPPLNELTEVPGEDKKKKKKKKDSVDTVAIKV | |
RUO | |
TMIE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
259236 | |
TMIE (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB6 | |
TMIE | |
Recombinant | |
wheat germ expression system |