Learn More
ACROBiosystems Human TIE2 Protein, Fc Tag
Marca:  ACROBiosystems TI2-H5255-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17206580
  • 2486.22€ / 1 miligramo
Fecha estimada de envío: 24-10-2022
para ver el stock
Descripción
Descripción
Tyrosine-protein kinase acts as cell-surface receptor for ANGPT1, ANGPT2 and ANGPT4, has anti-inflammatory effects by preventing the leakage of proinflammatory plasma proteins and leukocytes from blood vessels. Required for normal angiogenesis and heart development during embryogenesis, post-natal hematopoiesis. After birth, activates or inhibits angiogenesis, depending on the context. Inhibits angiogenesis and promotes vascular stability in quiescent vessels, where endothelial cells have tight contacts.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 107.5 kDa.
Especificaciones
Especificaciones
Powder
1 mg
Ala 23 - Leu 748
TIE2,Tie-2,TEK,VMCM, VMCM1,CD202b,Angiopoietin-1 receptor
Unconjugated
HEK293
Human
107.5 kDa
Human TIE2, Fc Tag (TI2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Leu 748 (Accession # Q02763-1).
-20°C
TIE2
Fc Tag
Other Recombinant Proteins