Learn More
Abnova™ Human THEA Partial ORF (AAH01517, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026027-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with acyl-CoA thioesterase activity towards medium (C12) and long-chain (C18) fatty acyl-CoA substrates. Expression of a similar murine protein in brown adipose tissue is induced by cold exposure and repressed by warmth. Expression of the mouse protein has been associated with obesity, with higher expression found in obesity-resistant mice compared with obesity-prone mice. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MADGEGYRNPTEVQMSQLVLPCHTNQRGELSVGQLLKWIDTTACLSAERHAGCPCVTASMDDIYFEHTISVGQVVNIKAKVNRAFNSSMEVGIQVASEDLCSEKQWNVCKEspecificaciones
AAH01517 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADGEGYRNPTEVQMSQLVLPCHTNQRGELSVGQLLKWIDTTACLSAERHAGCPCVTASMDDIYFEHTISVGQVVNIKAKVNRAFNSSMEVGIQVASEDLCSEKQWNVCK | |
RUO | |
ACOT11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26027 | |
THEA (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BFIT/BFIT1/BFIT2/KIAA0707/STARD14/THEA/THEM1 | |
ACOT11 | |
Recombinant | |
wheat germ expression system |