missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TGIF Full-length ORF (AAH00814, 1 a.a. - 272 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007050-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16101372

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain. Alternative splicing has been observed at this locus and eight variants, encoding four distinct isoforms, are described. [provided by RefSeq]

Sequence: MKGKKGIVAASGSETEDEDSMDIPLDLSSSAGSGKRRRRGNLPKESVQILRDWLYEHRYNAYPSEQEKALLSQQTHLSTLQVCNWFINARRRLLPDMLRKDGKDPNQFTISRRGAKISETSSVESVMGIKNFMPALEETPFHSCTAGPNPTLGRPLSPKPSSPGSVLARPSVICHTTVTALKDVPFSLCQSVGVGQNTDIQQIAAKNFTDTSLMYPEDTCKSGPSTNTQSGLFNTPPPTPPDLNQDFSGFQLLVDVALKRAAEMELQAKLTA
Especificaciones

Especificaciones

AAH00814
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.66kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPE4/MGC39747/MGC5066/TGIF
TGIF1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7050
TGIF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKGKKGIVAASGSETEDEDSMDIPLDLSSSAGSGKRRRRGNLPKESVQILRDWLYEHRYNAYPSEQEKALLSQQTHLSTLQVCNWFINARRRLLPDMLRKDGKDPNQFTISRRGAKISETSSVESVMGIKNFMPALEETPFHSCTAGPNPTLGRPLSPKPSSPGSVLARPSVICHTTVTALKDVPFSLCQSVGVGQNTDIQQIAAKNFTDTSLMYPEDTCKSGPSTNTQSGLFNTPPPTPPDLNQDFSGFQLLVDVALKRAAEMELQAKLTA
RUO
TGIF1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales