missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TEKT2 Full-length ORF (NP_055281.2, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00027285-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198722

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is expressed in the testis and its protein is localized to the flagella of the sperms, indicating that it may play a role in spermatogenesis. [provided by RefSeq]

Sequence: MATLSVKPSRRFQLPDWHTNSYLLSTNAQLQRDASHQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESAEQNLQAKNLPLDVAIECLTLRESRRDIDVVKDPVEDELHKEVEVIEATKKALQQKVSQAFEQLCLLQEVQQQLNSDHRGKMETLEIDRGCLSLNLRSPNISLKVDPTRVPDGSTTLQQWDDFSRFNKDRAEAEMKAATELREATALTIAETNNELEAQRVATEFAFRKRLREMEKVYSELKWQEKNTLEEIAELQEDIRHLEEDLRTKLLSLKLSHTRLEARTYRPNVELCRDQAQYGLTDEVHQLEATIAALKQKLAQAQDALDALCKHLARLQADIACKANSMLLDTKCMDTRRKLTVPAERFVPEVDTFTRTTNSTLSPLKSCQLELA
Especificaciones

Especificaciones

NP_055281.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TEKTB1/TEKTIN-T/h-tektin-t
TEKT2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
27285
TEKT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATLSVKPSRRFQLPDWHTNSYLLSTNAQLQRDASHQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESAEQNLQAKNLPLDVAIECLTLRESRRDIDVVKDPVEDELHKEVEVIEATKKALQQKVSQAFEQLCLLQEVQQQLNSDHRGKMETLEIDRGCLSLNLRSPNISLKVDPTRVPDGSTTLQQWDDFSRFNKDRAEAEMKAATELREATALTIAETNNELEAQRVATEFAFRKRLREMEKVYSELKWQEKNTLEEIAELQEDIRHLEEDLRTKLLSLKLSHTRLEARTYRPNVELCRDQAQYGLTDEVHQLEATIAALKQKLAQAQDALDALCKHLARLQADIACKANSMLLDTKCMDTRRKLTVPAERFVPEVDTFTRTTNSTLSPLKSCQLELA
RUO
TEKT2
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales