Learn More
Abnova™ Human TCEAL2 Full-length ORF (NP_525129.1, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00140597-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. [provided by RefSeq]
Sequence: MEKLFNENEGMPSNQGKIDNEEQPPHEGKPEVACILEDKKLENEGNTENTGKRVEEPLKDKEKPESAGKAKGEGKSERKGKSEMQGGSKTEGKPERGGRAEGEGEPDSEREPESEGEPESETRAAGKRPAEDDIPRKAKRKTNKGLAQYLKQYKEAIHDMNFSNEDMIREFDNMARVEDKRRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDTEDIPYVEspecificaciones
NP_525129.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MY0876G05/my048 | |
TCEAL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
140597 | |
TCEAL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKLFNENEGMPSNQGKIDNEEQPPHEGKPEVACILEDKKLENEGNTENTGKRVEEPLKDKEKPESAGKAKGEGKSERKGKSEMQGGSKTEGKPERGGRAEGEGEPDSEREPESEGEPESETRAAGKRPAEDDIPRKAKRKTNKGLAQYLKQYKEAIHDMNFSNEDMIREFDNMARVEDKRRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDTEDIPYV | |
RUO | |
TCEAL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |