Learn More
Abnova™ Human TCEA2 Partial ORF (NP_003186.1, 81 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006919-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is found in the nucleus, where it functions as an SII class transcription elongation factor. Elongation factors in this class are responsible for releasing RNA polymerase II ternary complexes from transcriptional arrest at template-encoded arresting sites. The encoded protein has been shown to interact with general transcription factor IIB, a basal transcription factor. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DASDAKARERGRGMPLPTSSRDASEAPDPSRKRPELPRAPSTPRITTFPPVPVTCDAVRNKCREMLTAALQTDHDHVAIGADCERLSAQIEECIFRDVGNEspecificaciones
NP_003186.1 | |
Liquid | |
6919 | |
TCEA2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TFIIS | |
TCEA2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DASDAKARERGRGMPLPTSSRDASEAPDPSRKRPELPRAPSTPRITTFPPVPVTCDAVRNKCREMLTAALQTDHDHVAIGADCERLSAQIEECIFRDVGN | |
RUO | |
TCEA2 | |
Wheat Germ (in vitro) | |
GST |