Learn More
Abnova™ Human TBCD Full-length ORF (AAH39654.1, 1 a.a. - 623 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006904-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state. [provided by RefSeq]
Sequence: MKISHWDGVIRELAARALHNLAQQAPEFSATQVFPRLLSMTLSPDLHTRHGSILACAEVAYALYKLAAQENRPVTDHLDEQAVQGLKQIHQQLYDRQLYRGLGGQLMRQAVCVLIEKLSLSKMPFRGDTVIDGWQWLINDTLRHLHLISSHSRQQMKDAAVSALAALCSEYYMKEPGEADPAIQEELITQYLAELRNPEEMTRCGFSLALGALPGFLLKGRLQQVLTGLRAVTHTSPEDVSFAESRRDGLKAIARICQTVGVKAGAPDEAVCGENVSQIYCALLGCMDDYTTDSRGDVGTWVRKAAMTSLMDLTLLLARSQPELIEAHTCERIMCCVAQQASEKIDRFRAHAASVFLTLLHFDSPPIPHVPHRGELEKLFPRSDVASVNWSAPSQAFPRITQLLGLPTYRYHVLLGLVVSLGGLTESTIRHSTQSLFEYMKGIQSDPQALGSFSGTLLQIFEDNLLNERVSVPLLKTLDHVLTHGCFDIFTTEEDHPFAVKLLALCKKEIKNSKDIQKLLSGIAVFCGMVQFPGDVRRQALLQLCLLLCHRFPLIRKTTASQVYETLLTYSDVVGADVLDEVVTVLSDTAWDAELAVVREQRNRLCDLLGVPRPQLVPQPGACEspecificaciones
AAH39654.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0988 | |
TBCD | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6904 | |
TBCD (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKISHWDGVIRELAARALHNLAQQAPEFSATQVFPRLLSMTLSPDLHTRHGSILACAEVAYALYKLAAQENRPVTDHLDEQAVQGLKQIHQQLYDRQLYRGLGGQLMRQAVCVLIEKLSLSKMPFRGDTVIDGWQWLINDTLRHLHLISSHSRQQMKDAAVSALAALCSEYYMKEPGEADPAIQEELITQYLAELRNPEEMTRCGFSLALGALPGFLLKGRLQQVLTGLRAVTHTSPEDVSFAESRRDGLKAIARICQTVGVKAGAPDEAVCGENVSQIYCALLGCMDDYTTDSRGDVGTWVRKAAMTSLMDLTLLLARSQPELIEAHTCERIMCCVAQQASEKIDRFRAHAASVFLTLLHFDSPPIPHVPHRGELEKLFPRSDVASVNWSAPSQAFPRITQLLGLPTYRYHVLLGLVVSLGGLTESTIRHSTQSLFEYMKGIQSDPQALGSFSGTLLQIFEDNLLNERVSVPLLKTLDHVLTHGCFDIFTTEEDHPFAVKLLALCKKEIKNSKDIQKLLSGIAVFCGMVQFPGDVRRQALLQLCLLLCHRFPLIRKTTASQVYETLLTYSDVVGADVLDEVVTVLSDTAWDAELAVVREQRNRLCDLLGVPRPQLVPQPGAC | |
RUO | |
TBCD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |