missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAPBP Partial ORF (NP_003181, 197 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006892-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166265

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a transmembrane glycoprotein which mediates interaction between newly assembled major histocompatibility complex (MHC) class I molecules and the transporter associated with antigen processing (TAP), which is required for the transport of antigenic peptides across the endoplasmic reticulum membrane. This interaction is essential for optimal peptide loading on the MHC class I molecule. Up to four complexes of MHC class I and this protein may be bound to a single TAP molecule. This protein contains a C-terminal double-lysine motif (KKKAE) known to maintain membrane proteins in the endoplasmic reticulum. This gene lies within the major histocompatibility complex on chromosome 6. Alternative splicing results in three transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: PGPPPFGLEWRRQHLGKGHLLLAATPGLNGQMPAAQEGAVAFAAWDDDEPWGPWTGNGTFWLPTVQPFQEGTYLATIHLPYLQGQVTLELAVYKPPKVSLMPAT
Especificaciones

Especificaciones

NP_003181
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PGPPPFGLEWRRQHLGKGHLLLAATPGLNGQMPAAQEGAVAFAAWDDDEPWGPWTGNGTFWLPTVQPFQEGTYLATIHLPYLQGQVTLELAVYKPPKVSLMPAT
RUO
TAPBP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6892
TAPBP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NGS17/TAPA/TPN/TPSN/tapasin
TAPBP
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales