Learn More
Abnova™ Human TAF1L Partial ORF (NP_722516, 1532 a.a. - 1641 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00138474-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This locus is intronless, and apparently arose in the primate lineage from retrotransposition of the transcript from the multi-exon TAF1 locus on the X chromosome. The gene is expressed in male germ cells, and the product has been shown to function interchangeably with the TAF1 product. [provided by RefSeq]
Sequence: NIVTQKMMAVPDSWPFHHPVNKKFVPDYYKMIVNPVDLETIRKNISKHKYQSRESFLDDVNLILANSVKYNGPESQYTKTAQEIVNICYQTITEYDEHLTQLEKDICTAKEspecificaciones
NP_722516 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NIVTQKMMAVPDSWPFHHPVNKKFVPDYYKMIVNPVDLETIRKNISKHKYQSRESFLDDVNLILANSVKYNGPESQYTKTAQEIVNICYQTITEYDEHLTQLEKDICTAK | |
RUO | |
TAF1L | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
138474 | |
TAF1L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC134910|TAF2A2 | |
TAF1L | |
Yes |