Learn More
Abnova™ Human SVIL Partial ORF (NP_003165, 1679 a.a. - 1786 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_003165 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 6840 |
Peso molecular | 37.62kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16106145
|
Abnova™
H00006840-Q01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 31-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16116145
|
Abnova™
H00006840-Q01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 31-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Description
This gene encodes a bipartite protein with distinct amino- and carboxy-terminal domains. The amino-terminus contains nuclear localization signals and the carboxy-terminus contains numerous consecutive sequences with extensive similarity to proteins in the gelsolin family of actin-binding proteins, which cap, nucleate, and/or sever actin filaments. The gene product is tightly associated with both actin filaments and plasma membranes, suggesting a role as a high-affinity link between the actin cytoskeleton and the membrane. The encoded protein appears to aid in both myosin II assembly during cell spreading and disassembly of focal adhesions. Two transcript variants encoding different isoforms of supervillin have been described. [provided by RefSeq]
Sequence: LIHAGLEPLTFTNMFPSWEHREDIAEITEMDTEVSNQITLVEDVLAKLCKTIYPLADLLARPLPEGVDPLKLEIYLTDEDFEFALDMTRDEYNALPAWKQVNLKKAKGSpécification
NP_003165 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A17191 | |
SVIL | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6840 | |
SVIL (Human) Recombinant Protein (Q01) | |
LIHAGLEPLTFTNMFPSWEHREDIAEITEMDTEVSNQITLVEDVLAKLCKTIYPLADLLARPLPEGVDPLKLEIYLTDEDFEFALDMTRDEYNALPAWKQVNLKKAKG | |
RUO | |
SVIL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |