Learn More
Abnova™ Human SUZ12 Partial ORF (NP_056170.1, 640 a.a. - 739 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023512-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region. [provided by RefSeq]
Sequence: VENYGQKIIKKNLCRNFMLHLVSMHDFNLISIMSIDKAVTKLREMQQKLEKGESASPANEEITEEQNGTANGFSEINSKEKALETDSVSGVSKQSKKQKLEspecificaciones
NP_056170.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VENYGQKIIKKNLCRNFMLHLVSMHDFNLISIMSIDKAVTKLREMQQKLEKGESASPANEEITEEQNGTANGFSEINSKEKALETDSVSGVSKQSKKQKL | |
RUO | |
SUZ12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23512 | |
SUZ12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHET9/JJAZ1/KIAA0160 | |
SUZ12 | |
Recombinant | |
wheat germ expression system |