missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SUSD4 Full-length ORF (NP_001032252.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00055061-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191073

  • 467.00€ / 25 microgramos


Fecha estimada de envío
19-02-2021
Añadir a la cesta

Descripción

Descripción

Sequence: MYHGMNPSNGDGFLEQQQQQQQPQSPQRLLAVILWFQLALCFGPAQLTGGFDDLQVCADPGIPENGFRTPSGGVFFEGSVARFHCQDGFKLKGATKRLCLKHFNGTLGWIPSDNSICVQEDCRIPQIEDAEIHNKTYRHGEKLIITCHEGFKIRYPDLHNMVSLCRDDGTWNNLPICQGCLRPLASSNGYVNISELQTSFPVGTVISYRCFPGFKLDGSAYLECLQNLIWSSSPPRCLALEGGRPEHLFPVLYFPHIRLAAAVLYFCPVLKSSPTPAPTCSSTSTTTSLF
Especificaciones

Especificaciones

NP_001032252.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10052|PRO222
SUSD4
Yes
wheat germ expression system
Liquid
55061
SUSD4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYHGMNPSNGDGFLEQQQQQQQPQSPQRLLAVILWFQLALCFGPAQLTGGFDDLQVCADPGIPENGFRTPSGGVFFEGSVARFHCQDGFKLKGATKRLCLKHFNGTLGWIPSDNSICVQEDCRIPQIEDAEIHNKTYRHGEKLIITCHEGFKIRYPDLHNMVSLCRDDGTWNNLPICQGCLRPLASSNGYVNISELQTSFPVGTVISYRCFPGFKLDGSAYLECLQNLIWSSSPPRCLALEGGRPEHLFPVLYFPHIRLAAAVLYFCPVLKSSPTPAPTCSSTSTTTSLF
RUO
SUSD4
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos