missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SURF5 Partial ORF (NP_006743.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006837-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16106135

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MAQQRALPQSKETLLQSYNKRLKDDIKSIMDNFTEIIKTAKIEDETQVSRATQGEQDNYEMHVRAANIVRAGESLMKLVSDLKQFLILNDFPSVNEAIDQ
Especificaciones

Especificaciones

NP_006743.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQQRALPQSKETLLQSYNKRLKDDIKSIMDNFTEIIKTAKIEDETQVSRATQGEQDNYEMHVRAANIVRAGESLMKLVSDLKQFLILNDFPSVNEAIDQ
RUO
MED22
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6837
SURF5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MED24/MGC48682/SURF5
MED22
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales