missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STYK1 Full-length ORF (NP_060893.1, 1 a.a. - 422 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055359-P01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Receptor protein tyrosine kinases, like STYK1, play important roles in diverse cellular and developmental processes, such as cell proliferation, differentiation, and survival (Liu et al., 2004 [PubMed 15150103]).[supplied by OMIM]
Sequence: MGMTRMLLECSLSDKLCVIQEKQYEVIIVPTLLVTIFLILLGVILWLFIREQRTQQQRSGPQGIAPVPPPRDLSWEAGHGGNVALPLKETSVENFLGATTPALAKLQVPREQLSEVLEQICSGSCGPIFRANMNTGDPSKPKSVILKALKEPAGLHEVQDFLGRIQFHQYLGKHKNLVQLEGCCTEKLPLYMVLEDVAQGDLLGFLWTCRRDVMTMDGLLYDLTEKQVYHIGKQVLLALEFLQEKHLFHGDVAARNILMQSDLTAKLCGLGLAYEVYTRGAISSTQTIPLKWLAPERLLLRPASIRADVWSFGILLYEMVTLGAPPYPEVPPTSILEHLQRRKIMKRPSSCTHTMYSIMKSCWRWREADRPSPRELRLRLEAAIKTADDEAVLQVPELVVPELYAAVAGIRVESLFYNYSMLEspecificaciones
NP_060893.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761P1010/NOK/SuRTK106 | |
STYK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55359 | |
STYK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGMTRMLLECSLSDKLCVIQEKQYEVIIVPTLLVTIFLILLGVILWLFIREQRTQQQRSGPQGIAPVPPPRDLSWEAGHGGNVALPLKETSVENFLGATTPALAKLQVPREQLSEVLEQICSGSCGPIFRANMNTGDPSKPKSVILKALKEPAGLHEVQDFLGRIQFHQYLGKHKNLVQLEGCCTEKLPLYMVLEDVAQGDLLGFLWTCRRDVMTMDGLLYDLTEKQVYHIGKQVLLALEFLQEKHLFHGDVAARNILMQSDLTAKLCGLGLAYEVYTRGAISSTQTIPLKWLAPERLLLRPASIRADVWSFGILLYEMVTLGAPPYPEVPPTSILEHLQRRKIMKRPSSCTHTMYSIMKSCWRWREADRPSPRELRLRLEAAIKTADDEAVLQVPELVVPELYAAVAGIRVESLFYNYSML | |
RUO | |
STYK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |