missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SRFBP1 Full-length ORF (NP_689759.2, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00153443-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MAQPGTLNLNNEVVKMRKEVKRIRVLVIRKLVRSVGRLKSKKGTEDALLKNQRRAQRLLEEIHAMKELKPDIVTKSALGDDINFEKIFKKPDSTATERAIARLAVHPLLKKKIDVLKAAVQAFKEARQNVAEVESSKNASEDNHSENTLYSNDNGSNLQREATVISEQKVKETKILAKKPIHNSKEKIAKMEHGPKAVTIANSPSKPSEKDSVVSLESQKTPADPKLKTLSQTKKNKGSDSSLSGNSDGGEEFCEEEKEYFDDSTEERFYKQSSMSEDSDSGDDFFIGKVRRTRKKESSCHSSVKEQKPLEKVFLKEDTGETHGDTRNDKIKPSTETRKLESVFFHSLSGSKSSRRNFKEQAPKTRSLDFPQNEPQIKNQFNKKLSGRLENTKQQLQLPLHPSWEASRRRKEQQSNIAVFQGKKITFDDEspecificaciones
NP_689759.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BUD22/FLJ25286/P49/Rlb1/STRAP/p49/STRAP | |
SRFBP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
153443 | |
SRFBP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAQPGTLNLNNEVVKMRKEVKRIRVLVIRKLVRSVGRLKSKKGTEDALLKNQRRAQRLLEEIHAMKELKPDIVTKSALGDDINFEKIFKKPDSTATERAIARLAVHPLLKKKIDVLKAAVQAFKEARQNVAEVESSKNASEDNHSENTLYSNDNGSNLQREATVISEQKVKETKILAKKPIHNSKEKIAKMEHGPKAVTIANSPSKPSEKDSVVSLESQKTPADPKLKTLSQTKKNKGSDSSLSGNSDGGEEFCEEEKEYFDDSTEERFYKQSSMSEDSDSGDDFFIGKVRRTRKKESSCHSSVKEQKPLEKVFLKEDTGETHGDTRNDKIKPSTETRKLESVFFHSLSGSKSSRRNFKEQAPKTRSLDFPQNEPQIKNQFNKKLSGRLENTKQQLQLPLHPSWEASRRRKEQQSNIAVFQGKKITFDD | |
RUO | |
SRFBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |