missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPIB Full-length ORF (NP_003112.2, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006689-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16173118

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

SPI1 (MIM 165170) and SPIB are members of a subfamily of ETS (see ETS1; MIM 164720) transcription factors. ETS proteins share a conserved ETS domain that mediates specific DNA binding. SPIB and SPI1 bind to a purine-rich sequence, the PU box (5-prime-GAGGAA-3-prime).[supplied by OMIM]

Sequence: MLALEAAQLDGPHFSCLYPDGVFYDLDSCKHSSYPDSEGAPDSLWDWTVAPPVPATPYEAFDPAAAAFSHPQAAQLCYEPPTYSPAGNLELAPSLEAPGPGLPAYPTENFASQTLVPPAYAPYPSPVLSEEEDLPLDSPALEVSDSESDEALVAGPEGKGSEAGTRKKLRLYQFLLGLLTRGDMRECVWWVEPGAGVFQFSSKHKELLARRWGQQKGNRKRMTYQKLARALRNYAKTGEIRKVKRKLTYQFDSALLPAVRRA
Especificaciones

Especificaciones

NP_003112.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPI-B
SPIB
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6689
SPIB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLALEAAQLDGPHFSCLYPDGVFYDLDSCKHSSYPDSEGAPDSLWDWTVAPPVPATPYEAFDPAAAAFSHPQAAQLCYEPPTYSPAGNLELAPSLEAPGPGLPAYPTENFASQTLVPPAYAPYPSPVLSEEEDLPLDSPALEVSDSESDEALVAGPEGKGSEAGTRKKLRLYQFLLGLLTRGDMRECVWWVEPGAGVFQFSSKHKELLARRWGQQKGNRKRMTYQKLARALRNYAKTGEIRKVKRKLTYQFDSALLPAVRRA
RUO
SPIB
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales