missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPHK2 Full-length ORF (NP_064511.2, 1 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056848-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108826

  • 518.16€ / 25 microgramos
Envío estimado: 18-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sequence: MNGHLEAEEQQDQRPDQELTGSWGHGPRSTLVRAKAMAPPPPPLAASTPLLHGEFGSYPARGPRFALTLTSQALHIQRLRPKPEARPRGGLVPLAEVSGCCTLRSRSPSDSAAYFCIYTYPRGRRGARRRATRTFRADGAATYEENRAEAQRWATALTCLLRGLPLPGDGEITPDLLPRPPRLLLLVNPFGGRGLAWQWCKNHVLPMISEAGLSFNLIQTERQNHARELVQGLSLSEWDGIVTVSGDGLLHEVLNGLLDRPDWEEAVKMPVGILPCGSGNALAGAVNQHGGFEPALGLDLLLNCSLLLCRGGGHPLDLLSVTLASGSRCFSFLSVAWGFVSDVDIQSERFRALGSARFTLGTVLGLATLHTYRGRLSYLPATVEPASPTPAHSLPRAKSELTLTPDPAPPMAHSPLHRSVSDLPLPLPQPALASPGSPEPLPILSLNGGGPELAGDWGGAGDAPLSPDPLLSSPPGSPKAALHSPVSEGAPVIPPSSGLPLPTPDARVGASTCGPPDHLLPPLGTPLPPDWVTLEGDFVLMLAISPSHLGADLVAAPHARFDDGLVHLCWVRSGISRAALLRLFLAMERGSHFSLGCPQLGYAAARAFRLEPLTPRGVLTVDGEQVEYGPLQAQMHPGIGTLLTGPPGCPGREP
Especificaciones

Especificaciones

NP_064511.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPHK2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
56848
SPHK2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGHLEAEEQQDQRPDQELTGSWGHGPRSTLVRAKAMAPPPPPLAASTPLLHGEFGSYPARGPRFALTLTSQALHIQRLRPKPEARPRGGLVPLAEVSGCCTLRSRSPSDSAAYFCIYTYPRGRRGARRRATRTFRADGAATYEENRAEAQRWATALTCLLRGLPLPGDGEITPDLLPRPPRLLLLVNPFGGRGLAWQWCKNHVLPMISEAGLSFNLIQTERQNHARELVQGLSLSEWDGIVTVSGDGLLHEVLNGLLDRPDWEEAVKMPVGILPCGSGNALAGAVNQHGGFEPALGLDLLLNCSLLLCRGGGHPLDLLSVTLASGSRCFSFLSVAWGFVSDVDIQSERFRALGSARFTLGTVLGLATLHTYRGRLSYLPATVEPASPTPAHSLPRAKSELTLTPDPAPPMAHSPLHRSVSDLPLPLPQPALASPGSPEPLPILSLNGGGPELAGDWGGAGDAPLSPDPLLSSPPGSPKAALHSPVSEGAPVIPPSSGLPLPTPDARVGASTCGPPDHLLPPLGTPLPPDWVTLEGDFVLMLAISPSHLGADLVAAPHARFDDGLVHLCWVRSGISRAALLRLFLAMERGSHFSLGCPQLGYAAARAFRLEPLTPRGVLTVDGEQVEYGPLQAQMHPGIGTLLTGPPGCPGREP
RUO
SPHK2
Yes
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales