missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SP3 Partial ORF (NP_003102.1, 516 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006670-Q02.10ug

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16054755

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 05-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. [provided by RefSeq]

Sequence: GQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNL
Especificaciones

Especificaciones

NP_003102.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue
GQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNL
RUO
SP3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6670
SP3 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686O1631/SPR-2
SP3
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales