Learn More
Abnova™ Human SORBS3 Full-length ORF (NP_001018003.1, 1 a.a. - 329 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010174-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The SORBS3 gene encodes 2 isoforms vinexin-alpha and -beta that are vinculin (VCL; MIM 193065)-binding cytoskeletal proteins involved in focal adhesion and cell-cell adhesion (Kioka et al., 1999 [PubMed 9885244]).[supplied by OMIM]
Sequence: MADGGSPFLGRRDFVYPSSTRDPSASNGGGSPARREEKKRKAARLKFDFQAQSPKELTLQKGDIVYIHKEVDKNWLEGEHHGRLGIFPANYVEVLPADEIPKPIKPPTYQVLEYGEAVAQYTFKGDLEVELSFRKGEHICLIRKVNENWYEGRITGTGRQGIFPASYVQVSREPRLRLCDDGPQLPTSPRLTAAARSARHPSSPSALRSPADPIDLGGQTSPRRTGFSFPTQEPRPQTQNLGTPGPALSHSRGPSHPLDLGTSSPNTSQIHWTPYRAMYQYRPQNEDELELREGDRVDVMQQCDDGWFVGVSRRTQKFGTFPGNYVAPVEspecificaciones
NP_001018003.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SCAM-1/SCAM1/SH3D4/VINEXIN | |
SORBS3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10174 | |
SORBS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADGGSPFLGRRDFVYPSSTRDPSASNGGGSPARREEKKRKAARLKFDFQAQSPKELTLQKGDIVYIHKEVDKNWLEGEHHGRLGIFPANYVEVLPADEIPKPIKPPTYQVLEYGEAVAQYTFKGDLEVELSFRKGEHICLIRKVNENWYEGRITGTGRQGIFPASYVQVSREPRLRLCDDGPQLPTSPRLTAAARSARHPSSPSALRSPADPIDLGGQTSPRRTGFSFPTQEPRPQTQNLGTPGPALSHSRGPSHPLDLGTSSPNTSQIHWTPYRAMYQYRPQNEDELELREGDRVDVMQQCDDGWFVGVSRRTQKFGTFPGNYVAPV | |
RUO | |
SORBS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |