Learn More
Abnova™ Human SOCS5 Partial ORF (AAH32862, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009655-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS) family, also known as STAT-induced STAT inhibitor (SSI) protein family.
Sequence:MDKVGKMWNNFKYRCQNLFGHEGGSRSENVDMNSNRCLSVKEKNISIGDSTPQQQSSPLRENIALQLGLSPSKNSSRRNQNCATEIPQIVEISIEKDNDSCVTPGTRLAR
Especificaciones
AAH32862 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDKVGKMWNNFKYRCQNLFGHEGGSRSENVDMNSNRCLSVKEKNISIGDSTPQQQSSPLRENIALQLGLSPSKNSSRRNQNCATEIPQIVEISIEKDNDSCVTPGTRLAR | |
RUO | |
SOCS5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9655 | |
SOCS5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIS6/CISH6/Cish5/KIAA0671/SOCS-5 | |
SOCS5 | |
Recombinant | |
wheat germ expression system |