missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLMAP Partial ORF (NP_009090, 677 a.a. - 784 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007871-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: LHNSQKQSLELTSDLSILQMSRKELENQVGSLKEQHLRDSADLKTLLSKAENQAKDVQKEYEKTQTVLSELKLKFEMTEQEKQSITDELKQCKNNLKLLREKGNNKPWEspecificaciones
NP_009090 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LHNSQKQSLELTSDLSILQMSRKELENQVGSLKEQHLRDSADLKTLLSKAENQAKDVQKEYEKTQTVLSELKLKFEMTEQEKQSITDELKQCKNNLKLLREKGNNKPW | |
RUO | |
SLMAP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7871 | |
SLMAP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42206/KIAA1601/MGC138760/MGC138761/SLAP | |
SLMAP | |
Recombinant | |
wheat germ expression system |