Learn More
Abnova™ Human SLC23A2 Full-length ORF (AAH13112.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009962-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1. [provided by RefSeq]
Sequence: MMGIGKNTTSKSMEAGSSTEGKYEDEAKHPAFFTLPVVINGGATSSGEQDNEDTELMAIYTTENGIAEKSSLAETLDSTGSLDPQRSDMIYTIEDVPPWYLCIFLGLQHYLTCFSGTIAVPFLLADAMCVGYDQWATSQLIGTIFFCVGITTLLQTTFGCRLPLFQASAFAFLAPARAILSLDKWKCNTTDVSVANGTAELLHTEHIWYPRIREIQGAIIMSSLIEVVIGLLGLPGALLKYIGPLTITPTVALIGLSGFQAAGERAGKHWGIAMLTIFLVLLFSQYARNVKFPLPIYKSKKGWTAYKLQLFKMFPIILAILVSWLLCFIFTVTDVFPPDSTKYGFYARTDARQGVLLVAPWFKVPYPFQWGLPTVSAAGVIGMLSAVVASIIESIGDYYACARLSCAPPPPIHAINRYVPEKTSSEspecificaciones
AAH13112.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0238/NBTL1/SLC23A1/SVCT2/YSPL2 | |
SLC23A2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9962 | |
SLC23A2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMGIGKNTTSKSMEAGSSTEGKYEDEAKHPAFFTLPVVINGGATSSGEQDNEDTELMAIYTTENGIAEKSSLAETLDSTGSLDPQRSDMIYTIEDVPPWYLCIFLGLQHYLTCFSGTIAVPFLLADAMCVGYDQWATSQLIGTIFFCVGITTLLQTTFGCRLPLFQASAFAFLAPARAILSLDKWKCNTTDVSVANGTAELLHTEHIWYPRIREIQGAIIMSSLIEVVIGLLGLPGALLKYIGPLTITPTVALIGLSGFQAAGERAGKHWGIAMLTIFLVLLFSQYARNVKFPLPIYKSKKGWTAYKLQLFKMFPIILAILVSWLLCFIFTVTDVFPPDSTKYGFYARTDARQGVLLVAPWFKVPYPFQWGLPTVSAAGVIGMLSAVVASIIESIGDYYACARLSCAPPPPIHAINRYVPEKTSS | |
RUO | |
SLC23A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |