Learn More
Abnova™ Human SIL1 Partial ORF (NP_071909, 201 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00064374-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Sequence: EKIAALFDLEYYVHQMDNAQDLLSFGGLQVVINGLNSTEPLVKEYAAFVLGAAFSSNPKVQVEAIEGGALQKLLVILATEQPLTAKKKVLFALCSLLRHFPYAQRQFLKEspecificaciones
NP_071909 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKIAALFDLEYYVHQMDNAQDLLSFGGLQVVINGLNSTEPLVKEYAAFVLGAAFSSNPKVQVEAIEGGALQKLLVILATEQPLTAKKKVLFALCSLLRHFPYAQRQFLK | |
RUO | |
SIL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64374 | |
SIL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAP/MSS/ULG5 | |
SIL1 | |
Recombinant | |
wheat germ expression system |