missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SIL1 Partial ORF (NP_071909, 201 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_071909
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 64374
Peso molecular 37.73kDa
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Product Code Brand Cantidad Price Quantity & Availability  
16109626
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00064374-Q01.25UG
25 ug
508.00€
25 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16199616
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00064374-Q01.10UG
10 ug
335.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]

Sequence: EKIAALFDLEYYVHQMDNAQDLLSFGGLQVVINGLNSTEPLVKEYAAFVLGAAFSSNPKVQVEAIEGGALQKLLVILATEQPLTAKKKVLFALCSLLRHFPYAQRQFLK
Specifications

Specifications

NP_071909
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAP/MSS/ULG5
SIL1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
64374
SIL1 (Human) Recombinant Protein (Q01)
EKIAALFDLEYYVHQMDNAQDLLSFGGLQVVINGLNSTEPLVKEYAAFVLGAAFSSNPKVQVEAIEGGALQKLLVILATEQPLTAKKKVLFALCSLLRHFPYAQRQFLK
RUO
SIL1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates