missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SH2D1B Full-length ORF (NP_444512.2, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00117157-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16197393

  • 458.00€ / 25 microgramos


Fecha estimada de envío
22-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MDLPYYHGRLTKQDCETLLLKEGVDGNFLLRDSESIPGVLCLCVSFKNIVYTYRIFREKHGYYRIQTAEGSPKQVFPSLKELISKFEKPNQGMVVHLLKPIKRTSPSLRWRGLKLELETFVNSNSDYVDVLP
Especificaciones

Especificaciones

NP_444512.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EAT2
SH2D1B
Yes
wheat germ expression system
Liquid
117157
SH2D1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDLPYYHGRLTKQDCETLLLKEGVDGNFLLRDSESIPGVLCLCVSFKNIVYTYRIFREKHGYYRIQTAEGSPKQVFPSLKELISKFEKPNQGMVVHLLKPIKRTSPSLRWRGLKLELETFVNSNSDYVDVLP
RUO
SH2D1B
Wheat Germ (in vitro)
GST
Videos
Documentos