missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SFXN3 Partial ORF (AAH00124.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00081855-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MESKMGELPLDINIQEPRWDQSTFLGRARHFFTVTDPRNLLLSGAQLEASRNIVQNYRAGVVTPGITEDQLWRAKYVYDSAFHPDTGEKVVLIGRMSAQVEspecificaciones
AAH00124.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESKMGELPLDINIQEPRWDQSTFLGRARHFFTVTDPRNLLLSGAQLEASRNIVQNYRAGVVTPGITEDQLWRAKYVYDSAFHPDTGEKVVLIGRMSAQV | |
RUO | |
SFXN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
81855 | |
SFXN3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BA108L7.2/SFX3 | |
SFXN3 | |
Recombinant | |
wheat germ expression system |