Learn More
Abnova™ Human SERTAD3 Full-length ORF (NP_037500.2, 1 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00029946-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene was identified in a yeast two-hybrid assay employing the second subunit of human replication protein A as bait. It is localized to the nucleus and its expression is significantly higher in cancer cell lines compared to normal cell lines. This protein has also been shown to be a strong transcriptional co-activator. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGSEspecificaciones
NP_037500.2 | |
Liquid | |
29946 | |
SERTAD3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGS | |
RUO | |
SERTAD3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBT1 | |
SERTAD3 | |
Yes | |
wheat germ expression system |