missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SELK Partial ORF (NP_067060, 39 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_067060
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 58515
Peso molecular 31.57kDa
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16109336
Ver documentos Detalles de la promoción
Abnova™
H00058515-Q01.25UG
25 ug
508.00€
25 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16199326
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00058515-Q01.10UG
10 ug
335.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This selenoprotein is localized to the endoplasmic reticulum and is highly expressed in the heart, where it may function as an antioxidant. [provided by RefSeq]

Sequence: KTLLQQDVKKRRSYGNSSDSRYDDGRGPPGNPPRRMGRINHLRGPSPPPMAGG
Specifications

Specifications

NP_067060
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.57kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPC030/HSPC297/MGC17057
SELK
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
58515
SELK (Human) Recombinant Protein (Q01)
KTLLQQDVKKRRSYGNSSDSRYDDGRGPPGNPPRRMGRINHLRGPSPPPMAGG
RUO
SELK
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates