Learn More
Abnova™ Human SEC3L1 Partial ORF (NP_839955, 780 a.a. - 879 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055763-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: EEEVSYQLAFNKQELRKVIKEYPGKEVKKGLDNLYKKVDKHLCEEENLLQVVWHSMQDEFIRQYKHFEGLIARCYPGSGVTMEFTIQDILDYCSSIAQSHEspecificaciones
NP_839955 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEEVSYQLAFNKQELRKVIKEYPGKEVKKGLDNLYKKVDKHLCEEENLLQVVWHSMQDEFIRQYKHFEGLIARCYPGSGVTMEFTIQDILDYCSSIAQSH | |
RUO | |
EXOC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55763 | |
SEC3L1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BM-102/FLJ10893/SEC3/SEC3L1/SEC3P | |
EXOC1 | |
Recombinant | |
wheat germ expression system |