Learn More
Abnova™ Human SEC23IP Full-length ORF (NP_009121.1, 1 a.a. - 1000 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011196-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
COPII-coated vesicles are involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway. [provided by RefSeq]
Sequence: MAERKPNGGSGGASTSSSGTNLLFSSSATEFSFNVPFIPVTQASASPASLLLPGEDSTDVGEEDSFLGQTSIHTSAPQTFSYFSQVSSSSDPFGNIGQSPLTTAATSVGQSGFPKPLTALPFTTGSQDVSNAFSPSISKAQPGAPPSSLMGINSYLPSQPSSLPPSYFGNQPQGIPQPGYNPYRHTPGSSRANPYIAPPQLQQCQTPGPPAHPPPSGPPVQMYQMPPGSLPPVPSSVQSPAQQQVPARPGAPSVQVPSPFLLQNQYEPVQPHWFYCKEVEYKQLWMPFSVFDSLNLEEIYNSVQPDPESVVLGTDGGRYDVYLYDRIRKAAYWEEEPAEVRRCTWFYKGDTDSRFIPYTEEFSEKLEAEYKKAVTTNQWHRRLEFPSGETIVMHNPKVIVQFQPSSVPDEWGTTQDGQTRPRVVKRGIDDNLDEIPDGEMPQVDHLVFVVHGIGPVCDLRFRSIIECVDDFRVVSLKLLRTHFKKSLDDGKVSRVEFLPVHWHSSLGGDATGVDRNIKKITLPSIGRFRHFTNETLLDILFYNSPTYCQTIVEKVGMEINHLHALFMSRNPDFKGGVSVAGHSLGSLILFDILSNQKDLNLSKCPGPLAVANGVVKQLHFQEKQMPEEPKLTLDESYDLVVENKEVLTLQETLEALSLSEYFSTFEKEKIDMESLLMCTVDDLKEMGIPLGPRKKIANFVEHKAAKLKKAASEKKAVAATSTKGQEQSAQKTKDMASLPSESNEPKRKLPVGACVSSVCVNYESFEVGAGQVSVAYNSLDFEPEIFFALGSPIAMFLTIRGVDRIDENYSLPTCKGFFNIYHPLDPVAYRLEPMIVPDLDLKAVLIPHHKGRKRLHLELKESLSRMGSDLKQGFISSLKSAWQTLNEFARAHTSSTQLQEELEKVANQIKEEEEKQVVEAEKVVESPDFSKDEDYLGKVGMLNGGRRIDYVLQEKPIESFNEYLFALQSHLCYWESEDTALLLLKEIYRTMNISPEQPQHEspecificaciones
NP_009121.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
137.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MSTP053/P125 | |
SEC23IP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11196 | |
SEC23IP (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAERKPNGGSGGASTSSSGTNLLFSSSATEFSFNVPFIPVTQASASPASLLLPGEDSTDVGEEDSFLGQTSIHTSAPQTFSYFSQVSSSSDPFGNIGQSPLTTAATSVGQSGFPKPLTALPFTTGSQDVSNAFSPSISKAQPGAPPSSLMGINSYLPSQPSSLPPSYFGNQPQGIPQPGYNPYRHTPGSSRANPYIAPPQLQQCQTPGPPAHPPPSGPPVQMYQMPPGSLPPVPSSVQSPAQQQVPARPGAPSVQVPSPFLLQNQYEPVQPHWFYCKEVEYKQLWMPFSVFDSLNLEEIYNSVQPDPESVVLGTDGGRYDVYLYDRIRKAAYWEEEPAEVRRCTWFYKGDTDSRFIPYTEEFSEKLEAEYKKAVTTNQWHRRLEFPSGETIVMHNPKVIVQFQPSSVPDEWGTTQDGQTRPRVVKRGIDDNLDEIPDGEMPQVDHLVFVVHGIGPVCDLRFRSIIECVDDFRVVSLKLLRTHFKKSLDDGKVSRVEFLPVHWHSSLGGDATGVDRNIKKITLPSIGRFRHFTNETLLDILFYNSPTYCQTIVEKVGMEINHLHALFMSRNPDFKGGVSVAGHSLGSLILFDILSNQKDLNLSKCPGPLAVANGVVKQLHFQEKQMPEEPKLTLDESYDLVVENKEVLTLQETLEALSLSEYFSTFEKEKIDMESLLMCTVDDLKEMGIPLGPRKKIANFVEHKAAKLKKAASEKKAVAATSTKGQEQSAQKTKDMASLPSESNEPKRKLPVGACVSSVCVNYESFEVGAGQVSVAYNSLDFEPEIFFALGSPIAMFLTIRGVDRIDENYSLPTCKGFFNIYHPLDPVAYRLEPMIVPDLDLKAVLIPHHKGRKRLHLELKESLSRMGSDLKQGFISSLKSAWQTLNEFARAHTSSTQLQEELEKVANQIKEEEEKQVVEAEKVVESPDFSKDEDYLGKVGMLNGGRRIDYVLQEKPIESFNEYLFALQSHLCYWESEDTALLLLKEIYRTMNISPEQPQH | |
RUO | |
SEC23IP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |