missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SDC2 Full-length ORF (AAH49836.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006383-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16100632

  • 326.95€ / 10 microgramos
Envío estimado: 20-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-2 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-2 expression has been detected in several different tumor types. [provided by RefSeq]

Sequence: MRRAWILLTLGLVACVSAESRAELTSDKDMYLDNSSIEEASGVYPIDDDDYASASGSGADEDVESPELTTTRPLPKILLTSAAPKVETTTLNIQNKIPAQTKSPEETDKEKVHLSDSERKMDPAEEDTNVYTEKHSDSLFKRTEVLAAVIAGGVIGFLFAIFLILLLVYRMRKKDEGSYDLGERKPSSAAYQKAPTKEFYA
Especificaciones

Especificaciones

AAH49836.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPG/HSPG1/SYND2
SDC2
Yes
wheat germ expression system
Liquid
6383
SDC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRRAWILLTLGLVACVSAESRAELTSDKDMYLDNSSIEEASGVYPIDDDDYASASGSGADEDVESPELTTTRPLPKILLTSAAPKVETTTLNIQNKIPAQTKSPEETDKEKVHLSDSERKMDPAEEDTNVYTEKHSDSLFKRTEVLAAVIAGGVIGFLFAIFLILLLVYRMRKKDEGSYDLGERKPSSAAYQKAPTKEFYA
RUO
SDC2
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales