Learn More
Abnova™ Human SDC2 Full-length ORF (AAH49836.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
en la oferta
Especificaciones
Nombre | SDC2 (Human) Recombinant Protein (P01) |
---|---|
Recombinante | Yes |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Peso molecular | 48.6kDa |
Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Descripción
The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-2 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-2 expression has been detected in several different tumor types. [provided by RefSeq]
Sequence: MRRAWILLTLGLVACVSAESRAELTSDKDMYLDNSSIEEASGVYPIDDDDYASASGSGADEDVESPELTTTRPLPKILLTSAAPKVETTTLNIQNKIPAQTKSPEETDKEKVHLSDSERKMDPAEEDTNVYTEKHSDSLFKRTEVLAAVIAGGVIGFLFAIFLILLLVYRMRKKDEGSYDLGERKPSSAAYQKAPTKEFYAEspecificaciones
SDC2 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
48.6kDa |