Learn More
Abnova™ Human SCYL1 Partial ORF (AAH09967, 373 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057410-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a transcriptional regulator belonging to the SCY1-like family of kinase-like proteins. The protein has a divergent N-terminal kinase domain that is thought to be catalytically inactive, and can bind specific DNA sequences through its C-terminal domain. It activates transcription of the telomerase reverse transcriptase and DNA polymerase beta genes. The protein has been localized to the nucleus, and also to the cytoplasm and centrosomes during mitosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DHKSSKSPESDWSSWEAEGSWEQGWQEPSSQEPPPDGTRLASEYNWGGPESSDKGDPFATLSARPSTQDRSRLSWPGRSARSGGGRWRPNAPRGRWPRAPEspecificaciones
AAH09967 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DHKSSKSPESDWSSWEAEGSWEQGWQEPSSQEPPPDGTRLASEYNWGGPESSDKGDPFATLSARPSTQDRSRLSWPGRSARSGGGRWRPNAPRGRWPRAP | |
RUO | |
SCYL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57410 | |
SCYL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GKLP/HT019/MGC78454/NKTL/NTKL/P105/TAPK/TEIF/TRAP | |
SCYL1 | |
Recombinant | |
wheat germ expression system |