Learn More
Abnova™ Human SCFD2 Full-length ORF (NP_689753.2, 1 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00152579-P01.25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16168733
  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-05-2023
para ver el stock
Descripción
Descripción
Sequence: MSASGVLSFTQQGWEQVLAKVKRAVVYLDAACAESLHWGCGSTRLLEAVGGPDCHLREFEPDAIGGGAKQPKAVFVLSCLLKGRTVEILRDIICRSHFQYCVVVTTVSHAVHLTANHVPAAAAAEMEGQQPVFEQLEEKLCEWMGNMNYTAEVFHVPLLLAPVAPHFALTPAFASLFPLLPQDVHLLNSARPDKRKLGSLGDVDSTTLTPELLLQIRCLVSGLSSLCEHLGVREECFAVGSLSQVIAADLANYAPAKNRKKTAAGRASVVFVDRTLDLTGAVGHHGDNLVEKIISALPQLPGHTNDVMVNMIALTALHTEEENYNVVAPGCLSQSSDTTAKALWEALLNTKHKEAVMEVRRHLVEAASRENLPIKMSMGRVTPGQLMSYIQLFKNNLKALMNHCGLLQLGLATAQTLKHPQTAKWDNFLAFERLLLQSIGESAMSVVLNQLLPMIKPVTQRTNEDYSPEELLILLIYIYSVTGELTVDKDLCEAEEKVKKALAQVFCEESGLSPLLQKITDWDSSINLTFHKSKIAVDELFTSLRDIAGARSLLKQFKSVYVPGNHTHQASYKPLLKQVVEEIFHPERPDSVDIEHMSSGLTDLLKTGFSMFMKVSRPHPSDYPLLILFVVGGVTVSEVKMVKDLVASLKPGTQVIVLSTRLLKPLNIPELLFATDRLHPDLGF
Especificaciones
Especificaciones
NP_689753.2
Liquid
152579
SCFD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSASGVLSFTQQGWEQVLAKVKRAVVYLDAACAESLHWGCGSTRLLEAVGGPDCHLREFEPDAIGGGAKQPKAVFVLSCLLKGRTVEILRDIICRSHFQYCVVVTTVSHAVHLTANHVPAAAAAEMEGQQPVFEQLEEKLCEWMGNMNYTAEVFHVPLLLAPVAPHFALTPAFASLFPLLPQDVHLLNSARPDKRKLGSLGDVDSTTLTPELLLQIRCLVSGLSSLCEHLGVREECFAVGSLSQVIAADLANYAPAKNRKKTAAGRASVVFVDRTLDLTGAVGHHGDNLVEKIISALPQLPGHTNDVMVNMIALTALHTEEENYNVVAPGCLSQSSDTTAKALWEALLNTKHKEAVMEVRRHLVEAASRENLPIKMSMGRVTPGQLMSYIQLFKNNLKALMNHCGLLQLGLATAQTLKHPQTAKWDNFLAFERLLLQSIGESAMSVVLNQLLPMIKPVTQRTNEDYSPEELLILLIYIYSVTGELTVDKDLCEAEEKVKKALAQVFCEESGLSPLLQKITDWDSSINLTFHKSKIAVDELFTSLRDIAGARSLLKQFKSVYVPGNHTHQASYKPLLKQVVEEIFHPERPDSVDIEHMSSGLTDLLKTGFSMFMKVSRPHPSDYPLLILFVVGGVTVSEVKMVKDLVASLKPGTQVIVLSTRLLKPLNIPELLFATDRLHPDLGF
RUO
SCFD2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
101.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21060/FLJ39514/STXBP1L1
SCFD2
Yes