Learn More
Abnova™ Human SCARA3 Partial ORF (NP_057324, 316 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051435-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a macrophage scavenger receptor-like protein. This protein has been shown to deplete reactive oxygen species, and thus play an important role in protecting cells from oxidative stress. The expression of this gene is induced by oxidative stress. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: SFLDDHEENMHDLQYHTHYAQNRTVERFESLEGRMASHEIEIGTIFTNINATDNHVHSMLKYLDDVRLSCTLGFHTHAEELYYLNKSVSIMLGTTDLLREEspecificaciones
NP_057324 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SFLDDHEENMHDLQYHTHYAQNRTVERFESLEGRMASHEIEIGTIFTNINATDNHVHSMLKYLDDVRLSCTLGFHTHAEELYYLNKSVSIMLGTTDLLRE | |
RUO | |
SCARA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51435 | |
SCARA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APC7/CSR/CSR1/MSLR1/MSRL1 | |
SCARA3 | |
Recombinant | |
wheat germ expression system |