missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPS29 Partial ORF (NP_001023, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_001023
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 6235
Peso molecular 31.9kDa
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16174765
Ver documentos Detalles de la promoción
Abnova™
H00006235-Q01.25UG
25 ug
508.00€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16164765
Ver documentos Detalles de la promoción
Abnova™
H00006235-Q01.10UG
10 ug
335.00€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit and a member of the S14P family of ribosomal proteins. The protein, which contains a C2-C2 zinc finger-like domain that can bind to zinc, can enhance the tumor suppressor activity of Ras-related protein 1A (KREV1). It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: MGHQQLYWSHPRKFGQGSRSCRVCSNRHGLIRKYGLNMCRQCFRQYAKDIGFIKLD
Especificaciones

Especificaciones

NP_001023
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.9kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPS29
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6235
RPS29 (Human) Recombinant Protein (Q01)
MGHQQLYWSHPRKFGQGSRSCRVCSNRHGLIRKYGLNMCRQCFRQYAKDIGFIKLD
RUO
RPS29
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Product Certifications